2-Bromo-1-ethoxy-4-fluoro-3-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

52886-500MG 52886-1G 52886-5G 52886-25G
77328-828EA 163.77 USD
77328-828 77346-086 77365-152 77387-232
2-Bromo-1-ethoxy-4-fluoro-3-iodobenzene ≥97%
2-Bromo-1-ethoxy-4-fluoro-3-iodobenzene

Precaución: for Research Use Only

Formula: C₈H₇BrFIO
MW: 344.947 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564224
CAS Number: 2383558-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR