2-(6-Chloro-2-fluoro-3-methylphenyl)propan-2-ol ≥97%

Supplier: AOBChem USA

Warning

52867-1G 52867-5G 52867-25G
77342-016EA 111.24 USD
77342-016 77365-142 77376-774
2-(6-Chloro-2-fluoro-3-methylphenyl)propan-2-ol ≥97%
2-(6-Chloro-2-fluoro-3-methylphenyl)propan-2-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₂ClFO
MW: 202.653 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33404098
CAS Number: 1534953-20-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR