(6-Chloro-2-fluoro-3-methylphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

52862-1G 52862-5G 52862-25G
77341-742EA 103 USD
77341-742 77365-138 77376-772
(6-Chloro-2-fluoro-3-methylphenyl)(methyl)sulfane ≥97%
(6-Chloro-2-fluoro-3-methylphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈ClFS
MW: 190.666 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28752247
CAS Number: 1805455-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR