(4-Chloro-2-methoxy-5-methylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

52851-500MG 52851-1G 52851-5G
77339-724EA 620.06 USD
77339-724 77352-530 77376-768
(4-Chloro-2-methoxy-5-methylphenyl)methanol ≥97%
(4-Chloro-2-methoxy-5-methylphenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₁ClO₂
MW: 186.635 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09999873
CAS Number: 1378822-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR