2-Bromo-6-fluoro-3-isopropoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

52849-500MG 52849-1G 52849-5G 52849-25G
77328-816EA 214.24 USD
77328-816 77352-528 77365-134 77387-220
2-Bromo-6-fluoro-3-isopropoxybenzoic acid ≥97%
2-Bromo-6-fluoro-3-isopropoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀BrFO₃
MW: 277.087 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178637
CAS Number: 2643367-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR