2-Bromo-6-chloro-3-methoxybenzoic acid ≥95%

Supplier: AOBChem USA

Warning

52843-500MG 52843-1G 52843-5G 52843-25G
77328-814EA 257.5 USD
77328-814 77352-526 77365-132 77387-218
2-Bromo-6-chloro-3-methoxybenzoic acid ≥95%
2-Bromo-6-chloro-3-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆BrClO₃
Storage Temperature: Ambient
MDL Number: MFCD30829704
CAS Number: 2089651-19-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR