2,5-Dibromo-4-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

52832-500MG 52832-1G 52832-5G 52832-25G
77328-812EA 252.35 USD
77328-812 77352-522 77365-130 77387-216
2,5-Dibromo-4-methoxybenzoic acid ≥97%
2,5-Dibromo-4-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆Br₂O₃
MW: 309.939 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31812140
CAS Number: 343789-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR