Methyl 4-chloro-2-methoxy-5-methylbenzoate ≥97%
Supplier: AOBChem USA
|
Warning
|
52828-500MG
52828-1G
52828-5G
52828-25G
77328-808EA
283.25
USD
77328-808
77352-520
77365-126
77387-212
Methyl 4-chloro-2-methoxy-5-methylbenzoate ≥97%
Methyl 4-chloro-2-methoxy-5-methylbenzoate
Precaución: for Research Use Only
Formula:
C₁₀H₁₁ClO₃ MW: 214.646 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD31695764 CAS Number: 1368228-61-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...