4-Iodo-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

52822-500MG 52822-1G 52822-5G 52822-25G
77328-804EA 116.39 USD
77328-804 77343-422 77365-124 77376-762
4-Iodo-2-methylbenzaldehyde ≥97%
4-Iodo-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₇IO
Storage Temperature: Ambient
MDL Number: MFCD18397776
CAS Number: 100304-96-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR