3-Bromo-5-isopropyl-2-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

52801-500MG 52801-1G 52801-5G
77328-798EA 332.69 USD
77328-798 77352-512 77376-758
3-Bromo-5-isopropyl-2-methylbenzaldehyde ≥97%
3-Bromo-5-isopropyl-2-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₁H₁₃BrO
Storage Temperature: Ambient
MDL Number: MFCD34756937
CAS Number: 2807466-45-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR