2-Bromo-4-isopropyl-1-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

52797-500MG 52797-1G 52797-5G 52797-25G
77328-796EA 209.09 USD
77328-796 77352-510 77365-120 77387-208
2-Bromo-4-isopropyl-1-methylbenzene ≥95%
2-Bromo-4-isopropyl-1-methylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃Br
MW: 213.114 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00059463
CAS Number: 2437-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR