5-Bromo-[1,1'-biphenyl]-3-carboxylic acid ≥97%

Supplier: AOBChem USA

Warning

52796-500MG 52796-1G 52796-5G 52796-25G
77328-794EA 191.58 USD
77328-794 77352-508 77365-118 77387-206
5-Bromo-[1,1'-biphenyl]-3-carboxylic acid ≥97%
5-Bromo-[1,1'-biphenyl]-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₉BrO₂
MW: 277.113 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21907473
CAS Number: 1368909-08-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR