(5-Bromo-[1,1'-biphenyl]-3-yl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

52794-500MG 52794-1G 52794-5G 52794-25G
77328-792EA 95.79 USD
77328-792 77342-558 77365-116 77376-756
(5-Bromo-[1,1'-biphenyl]-3-yl)boronic acid ≥97%
(5-Bromo-[1,1'-biphenyl]-3-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₀BBrO₂
MW: 276.922 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30541792
CAS Number: 2081938-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR