(3-tert-Butylphenyl)methanol ≥95%

Supplier: AOBChem USA

Warning

Synonyms: (3-(tert-Butyl)phenyl)methanol

52792-500MG 52792-1G 52792-5G
77336-188EA 380.07 USD
77336-188 77352-506 77376-754
(3-tert-Butylphenyl)methanol ≥95%
(3-tert-Butylphenyl)methanol

Precaución: for Research Use Only

Formula: C₁₁H₁₆O
MW: 164.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11111045
CAS Number: 51503-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR