1,5-Dibromo-3-(tert-butyl)-2-ethoxybenzene ≥97%

Supplier: AOBChem USA

Warning

52780-500MG 52780-1G 52780-5G 52780-25G
77328-788EA 185.4 USD
77328-788 77347-354 77365-112 77387-202
1,5-Dibromo-3-(tert-butyl)-2-ethoxybenzene ≥97%
1,5-Dibromo-3-(tert-butyl)-2-ethoxybenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₆Br₂O
MW: 336.063 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564221
CAS Number: 2734775-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR