Methyl 2,3-dimethyl-4-(methylthio)benzoate ≥97%
Supplier: AOBChem USA
|
Warning
|
52774-500MG
52774-1G
52774-5G
77328-784EA
334.75
USD
77328-784
77352-500
77376-750
Methyl 2,3-dimethyl-4-(methylthio)benzoate ≥97%
Methyl 2,3-dimethyl-4-(methylthio)benzoate
Precaución: for Research Use Only
Formula:
C₁₁H₁₄O₂S MW: 210.293 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD34564220 CAS Number: 2734779-60-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...