2,3-Dimethyl-4-(methylthio)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

52773-500MG 52773-1G 52773-5G
77336-910EA 399.64 USD
77336-910 77352-498 77376-748
2,3-Dimethyl-4-(methylthio)benzoic acid ≥97%
2,3-Dimethyl-4-(methylthio)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD33404093
CAS Number: 118939-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR