(2,3-Dimethyl-4-(methylthio)phenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

52772-500MG 52772-1G 52772-5G
77337-150EA 408.91 USD
77337-150 77352-496 77376-746
(2,3-Dimethyl-4-(methylthio)phenyl)methanol ≥97%
(2,3-Dimethyl-4-(methylthio)phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₀H₁₄OS
MW: 182.283 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34756934
CAS Number: 2755716-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR