2,3-Dimethyl-4-(methylthio)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

52771-500MG 52771-1G 52771-5G
77328-782EA 318.27 USD
77328-782 77352-494 77376-744
2,3-Dimethyl-4-(methylthio)benzaldehyde ≥97%
2,3-Dimethyl-4-(methylthio)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₂OS
MW: 180.267 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33404092
CAS Number: 2635937-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR