6-(Methylthio)picolinic acid ≥97%

Supplier: AOBChem USA

Warning

52757-500MG 52757-1G 52757-5G 52757-25G
77328-778EA 250.29 USD
77328-778 77352-490 77365-110 77387-198
6-(Methylthio)picolinic acid ≥97%
6-(Methylthio)picolinic acid

Precaución: for Research Use Only

Formula: C₇H₇NO₂S
Storage Temperature: Ambient
MDL Number: MFCD01860220
CAS Number: 74470-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR