(3-Fluoro-4-iodophenyl)(methyl)sulfane ≥95%

Supplier: AOBChem USA

Warning

52748-500MG 52748-1G 52748-5G 52748-25G
77328-776EA 176.13 USD
77328-776 77346-528 77365-108 77387-196
(3-Fluoro-4-iodophenyl)(methyl)sulfane ≥95%
(3-Fluoro-4-iodophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₇H₆FIS
Storage Temperature: Ambient
MDL Number: MFCD22489285
CAS Number: 1402705-21-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR