1,4-Dichloro-2-fluoro-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

52745-500MG 52745-1G 52745-5G 52745-25G
77328-772EA 190.55 USD
77328-772 77352-488 77365-104 77387-192
1,4-Dichloro-2-fluoro-3-methylbenzene ≥97%
1,4-Dichloro-2-fluoro-3-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₅Cl₂F
MW: 179.019 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28786358
CAS Number: 1804881-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR