(3,6-Dichloro-2-fluorophenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

52744-500MG 52744-1G 52744-5G 52744-25G
77328-770EA 138.02 USD
77328-770 77344-654 77365-102 77387-190
(3,6-Dichloro-2-fluorophenyl)(methyl)sulfane ≥97%
(3,6-Dichloro-2-fluorophenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₇H₅Cl₂FS
MW: 211.084 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28786348
CAS Number: 1804421-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR