2-Fluoro-3-methoxy-4-phenylpyridine ≥97%

Supplier: AOBChem USA

Warning

52743-500MG 52743-1G 52743-5G
77328-768EA 318.27 USD
77328-768 77352-486 77376-740
2-Fluoro-3-methoxy-4-phenylpyridine ≥97%
2-Fluoro-3-methoxy-4-phenylpyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₀FNO
MW: 203.212 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28797115
CAS Number: 1806393-08-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR