4'-Bromo-3,4-dimethoxy-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

52732-500MG 52732-1G 52732-5G 52732-25G
77328-762EA 282.22 USD
77328-762 77352-482 77365-096 77387-186
4'-Bromo-3,4-dimethoxy-1,1'-biphenyl ≥95%
4'-Bromo-3,4-dimethoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃BrO₂
MW: 293.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26635067
CAS Number: 74447-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR