Ethyl 3-cyano-4-methylbenzoate ≥97%

Supplier: AOBChem USA

Warning

52730-500MG 52730-1G 52730-5G 52730-25G
77328-760EA 145.23 USD
77328-760 77345-130 77365-094 77387-184
Ethyl 3-cyano-4-methylbenzoate ≥97%
Ethyl 3-cyano-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₂
MW: 189.211 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18396256
CAS Number: 96259-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR