4,6-dimethylisophthalaldehyde ≥97%

Supplier: AOBChem USA

Warning

52695-500MG 52695-1G 52695-5G
77339-016EA 544.87 USD
77339-016 77352-476 77376-734
4,6-dimethylisophthalaldehyde ≥97%
4,6-dimethylisophthalaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₂
Storage Temperature: Ambient
MDL Number: MFCD24673937
CAS Number: 25445-41-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR