1-(2-Fluoro-4-iodophenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

52678-500MG 52678-1G 52678-5G 52678-25G
77328-752EA 178.19 USD
77328-752 77346-608 77365-086 77387-176
1-(2-Fluoro-4-iodophenyl)ethanone ≥97%
1-(2-Fluoro-4-iodophenyl)ethanone

Precaución: for Research Use Only

Formula: C₈H₆FIO
Storage Temperature: Ambient
MDL Number: MFCD15527699
CAS Number: 249291-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR