(2,6-Difluoro-3-iodophenyl)trimethylsilane ≥97%

Supplier: AOBChem USA

Warning

52672-1G 52672-5G 52672-25G
77341-278EA 80.34 USD
77341-278 77365-084 77376-732
(2,6-Difluoro-3-iodophenyl)trimethylsilane ≥97%
(2,6-Difluoro-3-iodophenyl)trimethylsilane

Precaución: for Research Use Only

Formula: C₉H₁₁F₂ISi
MW: 312.17 g/mol
Storage Temperature: Ambient
MDL Number: MFCD30178311
CAS Number: 746630-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR