5-Methoxy-2,4-dimethylbenzoic acid ≥97%

Supplier: AOBChem USA

Warning

52623-500MG 52623-1G 52623-5G
77340-128EA 770.44 USD
77340-128 77352-464 77376-726
5-Methoxy-2,4-dimethylbenzoic acid ≥97%
5-Methoxy-2,4-dimethylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD20641137
CAS Number: 32644-90-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR