1-Bromo-5-methoxy-2,4-dimethylbenzene ≥95%

Supplier: AOBChem USA

Warning

52621-500MG 52621-1G 52621-5G
77338-658EA 507.79 USD
77338-658 77352-462 77376-724
1-Bromo-5-methoxy-2,4-dimethylbenzene ≥95%
1-Bromo-5-methoxy-2,4-dimethylbenzene

Precaución: for Research Use Only

Formula: C₉H₁₁BrO
Storage Temperature: Ambient
MDL Number: MFCD28738462
CAS Number: 77771-60-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR