Ethyl 3,4-difluoro-2-isopropoxybenzoate ≥95%

Supplier: AOBChem USA

Warning

52617-500MG 52617-1G 52617-5G
77336-810EA 394.49 USD
77336-810 77352-460 77376-722
Ethyl 3,4-difluoro-2-isopropoxybenzoate ≥95%
Ethyl 3,4-difluoro-2-isopropoxybenzoate

Precaución: for Research Use Only

Formula: C₁₂H₁₄F₂O₃
MW: 244.235 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530179
CAS Number: 2109969-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR