(2-Fluoro-3-iodo-6-methoxyphenyl)(methyl)sulfane ≥97%

Supplier: AOBChem USA

Warning

52609-500MG 52609-1G 52609-5G 52609-25G
77328-746EA 177.16 USD
77328-746 77346-598 77365-078 77387-170
(2-Fluoro-3-iodo-6-methoxyphenyl)(methyl)sulfane ≥97%
(2-Fluoro-3-iodo-6-methoxyphenyl)(methyl)sulfane

Precaución: for Research Use Only

Formula: C₈H₈FIOS
Storage Temperature: Ambient
MDL Number: MFCD33404084
CAS Number: 2635937-43-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR