2-Ethoxy-5-(trifluoromethoxy)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

52604-500MG 52604-1G 52604-5G
77336-620EA 390.37 USD
77336-620 77352-454 77376-716
2-Ethoxy-5-(trifluoromethoxy)benzaldehyde ≥97%
2-Ethoxy-5-(trifluoromethoxy)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₉F₃O₃
MW: 234.172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27944893
CAS Number: 145742-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR