4-(Benzyloxy)-2-fluoro-5-methylbenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

52600-500MG 52600-1G 52600-5G
77338-868EA 527.36 USD
77338-868 77352-450 77376-712
4-(Benzyloxy)-2-fluoro-5-methylbenzaldehyde ≥97%
4-(Benzyloxy)-2-fluoro-5-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₁₃FO₂
Storage Temperature: Ambient
MDL Number: MFCD34178635
CAS Number: 2643368-57-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR