2,2-Difluoro-1-(2-methoxyphenyl)ethanone ≥95%

Supplier: AOBChem USA

Warning

52573-500MG 52573-1G 52573-5G 52573-25G
77328-744EA 235.87 USD
77328-744 77352-442 77365-076 77387-168
2,2-Difluoro-1-(2-methoxyphenyl)ethanone ≥95%
2,2-Difluoro-1-(2-methoxyphenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₈F₂O₂
MW: 186.155 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16618124
CAS Number: 1352225-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR