5-Bromo-3-chloro-2-fluoro-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

52559-500MG 52559-1G 52559-5G
77337-422EA 414.06 USD
77337-422 77352-434 77376-702
5-Bromo-3-chloro-2-fluoro-1,1'-biphenyl ≥95%
5-Bromo-3-chloro-2-fluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₇BrClF
MW: 285.539 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178634
CAS Number: 2643367-59-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR