2-Chloro-3-(methoxymethyl)-4-phenylpyridine ≥97%

Supplier: AOBChem USA

Warning

52556-500MG 52556-1G 52556-5G
77339-354EA 581.95 USD
77339-354 77352-432 77376-700
2-Chloro-3-(methoxymethyl)-4-phenylpyridine ≥97%
2-Chloro-3-(methoxymethyl)-4-phenylpyridine

Precaución: for Research Use Only

Formula: C₁₃H₁₂ClNO
MW: 233.693 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178633
CAS Number: 2643368-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR