4-(2-Chloro-3-(methoxymethyl)pyridin-4-yl)benzaldehyde ≥97%

Supplier: AOBChem USA

Warning

52553-500MG 52553-1G 52553-5G
77339-580EA 600.49 USD
77339-580 77352-428 77376-696
4-(2-Chloro-3-(methoxymethyl)pyridin-4-yl)benzaldehyde ≥97%
4-(2-Chloro-3-(methoxymethyl)pyridin-4-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₂ClNO₂
MW: 261.704 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34178630
CAS Number: 2643368-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR