1,3-Dibromo-4-methoxy-2-methylbenzene ≥95%

Supplier: AOBChem USA

Warning

52551-500MG 52551-1G 52551-5G 52551-25G
77328-736EA 234.84 USD
77328-736 77352-426 77365-070 77387-162
1,3-Dibromo-4-methoxy-2-methylbenzene ≥95%
1,3-Dibromo-4-methoxy-2-methylbenzene

Precaución: for Research Use Only

Formula: C₈H₈Br₂O
MW: 279.957 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33404077
CAS Number: 688744-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR