2-Chloro-5-ethyl-1,1'-biphenyl ≥95%

Supplier: AOBChem USA

Warning

52544-500MG 52544-1G
77340-424EA 1037.21 USD
77340-424 77352-424
2-Chloro-5-ethyl-1,1'-biphenyl ≥95%
2-Chloro-5-ethyl-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₄H₁₃Cl
Storage Temperature: Ambient
CAS Number: 2918964-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR