4-Ethyl-1,2-diiodobenzene ≥97%

Supplier: AOBChem USA

Warning

52542-500MG 52542-1G 52542-5G 52542-25G
77328-732EA 259.56 USD
77328-732 77352-420 77365-066 77387-160
4-Ethyl-1,2-diiodobenzene ≥97%
4-Ethyl-1,2-diiodobenzene

Precaución: for Research Use Only

Formula: C₈H₈I₂
MW: 357.958 g/mol
Storage Temperature: Ambient
MDL Number: MFCD29095355
CAS Number: 1369792-52-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR