(2-Bromo-4-ethylphenyl)methanol ≥97%

Supplier: AOBChem USA

Warning

52528-500MG 52528-1G 52528-5G
77338-406EA 486.16 USD
77338-406 77352-414 77376-690
(2-Bromo-4-ethylphenyl)methanol ≥97%
(2-Bromo-4-ethylphenyl)methanol

Precaución: for Research Use Only

Formula: C₉H₁₁BrO
Storage Temperature: Ambient
MDL Number: MFCD22491785
CAS Number: 1427362-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR