1,2-Dichloro-4-fluoro-5-iodo-3-methylbenzene ≥97%

Supplier: AOBChem USA

Warning

52520-500MG 52520-1G 52520-5G 52520-25G
77328-724EA 184.37 USD
77328-724 77347-156 77365-058 77387-152
1,2-Dichloro-4-fluoro-5-iodo-3-methylbenzene ≥97%
1,2-Dichloro-4-fluoro-5-iodo-3-methylbenzene

Precaución: for Research Use Only

Formula: C₇H₄Cl₂FI
Storage Temperature: Ambient
MDL Number: MFCD33404073
CAS Number: 2635937-13-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR