3-Bromo-6-chloro-2-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

52503-500MG 52503-1G 52503-5G
77340-388EA 986.74 USD
77340-388 77352-398 77376-678
3-Bromo-6-chloro-2-methoxybenzoic acid ≥97%
3-Bromo-6-chloro-2-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₈H₆BrClO₃
Storage Temperature: Ambient
MDL Number: MFCD28154797
CAS Number: 1823566-09-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR