4-Isopropoxy-2-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

52493-500MG 52493-1G 52493-5G 52493-25G
77328-712EA 189.52 USD
77328-712 77352-396 77365-054 77387-142
4-Isopropoxy-2-methoxybenzoic acid ≥97%
4-Isopropoxy-2-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₄
MW: 210.226 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24483451
CAS Number: 79128-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR