4-Ethoxy-2-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

52486-500MG 52486-1G 52486-5G 52486-25G
77328-708EA 202.91 USD
77328-708 77352-392 77365-050 77387-138
4-Ethoxy-2-methoxybenzoic acid ≥97%
4-Ethoxy-2-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₄
Storage Temperature: Ambient
MDL Number: MFCD20638910
CAS Number: 55744-85-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR