(2,3-Difluoro-1,4-phenylene)bis(methylsulfane) ≥97%

Supplier: AOBChem USA

Warning

52478-500MG 52478-1G 52478-5G
77328-706EA 324.45 USD
77328-706 77352-388 77376-674
(2,3-Difluoro-1,4-phenylene)bis(methylsulfane) ≥97%
(2,3-Difluoro-1,4-phenylene)bis(methylsulfane)

Precaución: for Research Use Only

Formula: C₈H₈F₂S₂
MW: 206.276 g/mol
Storage Temperature: Ambient
MDL Number: MFCD33404070
CAS Number: 2390118-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR