1-(2,3-Difluoro-4-(methylthio)phenyl)ethanone ≥97%

Supplier: AOBChem USA

Warning

52476-500MG 52476-1G 52476-5G
77338-184EA 468.65 USD
77338-184 77352-386 77376-672
1-(2,3-Difluoro-4-(methylthio)phenyl)ethanone ≥97%
1-(2,3-Difluoro-4-(methylthio)phenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₈F₂OS
Storage Temperature: Ambient
MDL Number: MFCD33404069
CAS Number: 1896409-47-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR