2-(2,3-Difluoro-4-(methylthio)phenyl)propan-2-ol ≥97%

Supplier: AOBChem USA

Warning

52475-500MG 52475-1G 52475-5G 52475-25G
77328-704EA 279.13 USD
77328-704 77352-384 77365-048 77387-136
2-(2,3-Difluoro-4-(methylthio)phenyl)propan-2-ol ≥97%
2-(2,3-Difluoro-4-(methylthio)phenyl)propan-2-ol

Precaución: for Research Use Only

Formula: C₁₀H₁₂F₂OS
Storage Temperature: Ambient
MDL Number: MFCD33404068
CAS Number: 1895582-69-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR